Learn More
Medchemexpress LLC KRAS G12C inhibitor 15 | 2349393-21-9 | 98.5% | 498.91 g/mol | C25H21ClF2N4O3 | 50MG
SDP
Catalog No. 5000238308
Encompass_Preferred
KRAS G12C inhibitor 15 is a small-molecule research compound reported to selectively inhibit the oncogenic KRAS G12C mutant. Described in patent WO2019110751A1 (compound 22), it demonstrates subnanomolar-nanomolar biochemical potency and is supplied as a high-purity solid for preclinical and biochemical studies.
  • Potent KRAS G12C inhibitor with reported IC50 ≈ 5 nM.
  • Registered CAS number 2349393-21-9 for unambiguous identification.
  • High purity suitable for research applications (manufacturer listed ~98.5%).
  • Molecular formula C25H21ClF2N4O3 and molecular weight 498.91 g/mol.
  • Intended for biochemical assays, cellular studies, and tool-compound research.
  • Supplied as a solid; store at -20 °C in a dry, inert atmosphere for stability.
  • Available in multiple small pack sizes to support dose-range experiments.
Catalog No. 50-002-38308 Supplier Medchemexpress LLC Supplier No. HY12587350MG
Add to Cart
Add to Cart
Request a Quote
KRAS G12C inhibitor 15 is a small-molecule research compound reported to selectively inhibit the oncogenic KRAS G12C mutant. Described in patent WO2019110751A1 (compound 22), it demonstrates subnanomolar-nanomolar biochemical potency and is supplied as a high-purity solid for preclinical and biochemical studies.
  • Potent KRAS G12C inhibitor with reported IC50 ≈ 5 nM.
  • Registered CAS number 2349393-21-9 for unambiguous identification.
  • High purity suitable for research applications (manufacturer listed ~98.5%).
  • Molecular formula C25H21ClF2N4O3 and molecular weight 498.91 g/mol.
  • Intended for biochemical assays, cellular studies, and tool-compound research.
  • Supplied as a solid; store at -20 °C in a dry, inert atmosphere for stability.
  • Available in multiple small pack sizes to support dose-range experiments.