Learn More
AURORA BIOLABS LLC Kras G12D Binding Assay, 384 rxn
Catalog No. NC3786919
Encompass
The Aurora Biolabs KRAS G12D c-RAF Binding Assay Kit is a TR-FRET based assay used to quantify binding between active KRAS G12D and the Ras Binding Domain (RBD) of c-RAF. The KRAS protein is preloaded with GTP and recognized by a terbium-labeled anti-Tag2 antibody (HTRF donor), while the c-RAF-RBD is labeled with a fluorescent acceptor. Upon binding, FRET occurs and emission at 665 nm is measured. This assay provides a robust method for screening inhibitors that block KRAS effector interactions. Ideal for oncology research and drug discovery, each kit contains reagents for 384 reactions.
Catalog No. NC3786919 Supplier AURORA BIOLABS LLC Supplier No. 57274123BK384
The Aurora Biolabs KRAS G12D c-RAF Binding Assay Kit is a TR-FRET based assay used to quantify binding between active KRAS G12D and the Ras Binding Domain (RBD) of c-RAF. The KRAS protein is preloaded with GTP and recognized by a terbium-labeled anti-Tag2 antibody (HTRF donor), while the c-RAF-RBD is labeled with a fluorescent acceptor. Upon binding, FRET occurs and emission at 665 nm is measured. This assay provides a robust method for screening inhibitors that block KRAS effector interactions. Ideal for oncology research and drug discovery, each kit contains reagents for 384 reactions.