Learn More
AURORA BIOLABS LLC Kras G12D, GDP Loaded, 100 μg
Catalog No. NC3787932
Encompass
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12D substitution mutation favors the activated, GTP-bound state of the protein.
Catalog No. NC3787932 Supplier AURORA BIOLABS LLC Supplier No. 57274123G G100
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12D substitution mutation favors the activated, GTP-bound state of the protein.