Learn More
Medchemexpress LLC KRAS G12D-IN-31 | 3024060-23-6 | 98.5% | 698.80 | 1 MG
SDP
Catalog No. 5000392948
Encompass_Preferred
KRAS G12D-IN-31 is a potent inhibitor of KRAS G12D, demonstrating an IC50 of less than 100 nM. It effectively inhibits the proliferation of various RAS-dependent cells, including those with KRASG12C, KRAS G12D, KRASG12V, and KRASWT mutations. This compound is a valuable tool for research into conditions such as non-small cell lung cancer, gastric cancer, colon cancer, and malignant melanoma.
  • Potent KRAS G12D inhibitor
  • Inhibits proliferation of RAS-dependent cells
  • Suitable for research on non-small cell lung cancer
  • Suitable for research on gastric cancer
  • Suitable for research on colon cancer
  • Suitable for research on malignant melanoma
Catalog No. 50-003-92948 Supplier Medchemexpress LLC Supplier No. HY1635951MG
KRAS G12D-IN-31 is a potent inhibitor of KRAS G12D, demonstrating an IC50 of less than 100 nM. It effectively inhibits the proliferation of various RAS-dependent cells, including those with KRASG12C, KRAS G12D, KRASG12V, and KRASWT mutations. This compound is a valuable tool for research into conditions such as non-small cell lung cancer, gastric cancer, colon cancer, and malignant melanoma.
  • Potent KRAS G12D inhibitor
  • Inhibits proliferation of RAS-dependent cells
  • Suitable for research on non-small cell lung cancer
  • Suitable for research on gastric cancer
  • Suitable for research on colon cancer
  • Suitable for research on malignant melanoma