Learn More
Medchemexpress LLC Kras G12d Inhibitor 14 | 2765254-39-3 | 98.88% | C20H19F3N4OS | 1 ML
SDP
Catalog No. 5000356674
Encompass_Preferred
KRAS G12D inhibitor 14 is a potent KRAS G12D inhibitor with a KD of 33 nM for binding to KRAS G12D protein. It decreases the active form of KRAS G12D (KRAS G12D-GTP) but not KRAS G13D. For research use only, not for sale to patients.
  • Binds to KRAS G12D protein with a KD of 33 nM.
  • Decreases the active form of KRAS G12D (KRAS G12D-GTP) but does not affect KRAS G13D.
  • Shows antiproliferative activity with an IC50 of 2.1 μM against KRAS G12D-mutated cells.
  • Causes efficient apoptosis in SW1990 and CT26 cancer cells.
  • Down-regulates phosphorylated Raf and Erk in CT26 and SW1990 cancer cell lines.
  • Exhibits significant antitumor efficacy in a CT26 tumor model with a tumor growth inhibition (TGI) of 42% or 53% at doses of 40 mg/kg or 60 mg/kg (i.p.), without apparent toxicity.
Catalog No. 50-003-56674 Supplier Medchemexpress LLC Supplier No. HY14466110MM/1ML
KRAS G12D inhibitor 14 is a potent KRAS G12D inhibitor with a KD of 33 nM for binding to KRAS G12D protein. It decreases the active form of KRAS G12D (KRAS G12D-GTP) but not KRAS G13D. For research use only, not for sale to patients.
  • Binds to KRAS G12D protein with a KD of 33 nM.
  • Decreases the active form of KRAS G12D (KRAS G12D-GTP) but does not affect KRAS G13D.
  • Shows antiproliferative activity with an IC50 of 2.1 μM against KRAS G12D-mutated cells.
  • Causes efficient apoptosis in SW1990 and CT26 cancer cells.
  • Down-regulates phosphorylated Raf and Erk in CT26 and SW1990 cancer cell lines.
  • Exhibits significant antitumor efficacy in a CT26 tumor model with a tumor growth inhibition (TGI) of 42% or 53% at doses of 40 mg/kg or 60 mg/kg (i.p.), without apparent toxicity.