Learn More
AURORA BIOLABS LLC Kras G12R, GppNHp loaded, 100 μg
Catalog No. NC3787938
Encompass
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12R substitution mutation favors the activated, GTP-bound state of the protein.
Catalog No. NC3787938 Supplier AURORA BIOLABS LLC Supplier No. 57274127GGP100
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12R substitution mutation favors the activated, GTP-bound state of the protein.