Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRas G12R inhibitor 1 | C39H38ClF7N6O9 | 1 MG
SDP

Catalog No. 5000372149
Encompass_Preferred

KRas G12R inhibitor 1 is a covalent inhibitor that specifically targets the oncogenic mutant KRas G12R. It functions by exploiting the strong nucleophilicity of the mutant cysteine and irreversibly binding in the Switch II region of KRas, due to its α,β-diketoamide structure. This product can be used in research for K-Ras (G12R)-driven cancers, such as pancreatic ductal adenocarcinoma (PDAC).

  • Covalent inhibitor targeting KRas G12R mutant
  • Selectivity for the mutant arginine
  • Irreversibly binds in the Switch II region of KRas
  • Possesses an α,β-diketoamide structure
  • Potential for research in K-Ras (G12R)-driven cancers like pancreatic ductal adenocarcinoma (PDAC)
  • Purity: 98.40%
  • Appearance: Solid, white to off-white

Catalog No. 50-003-72149 Supplier Medchemexpress LLC Supplier No. HY1515231MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRas G12R inhibitor 1 is a covalent inhibitor that specifically targets the oncogenic mutant KRas G12R. It functions by exploiting the strong nucleophilicity of the mutant cysteine and irreversibly binding in the Switch II region of KRas, due to its α,β-diketoamide structure. This product can be used in research for K-Ras (G12R)-driven cancers, such as pancreatic ductal adenocarcinoma (PDAC).

  • Covalent inhibitor targeting KRas G12R mutant
  • Selectivity for the mutant arginine
  • Irreversibly binds in the Switch II region of KRas
  • Possesses an α,β-diketoamide structure
  • Potential for research in K-Ras (G12R)-driven cancers like pancreatic ductal adenocarcinoma (PDAC)
  • Purity: 98.40%
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.