Learn More
AURORA BIOLABS LLC Kras G13D, GppNHp loaded, 100 μg
Catalog No. NC3787942
Encompass
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. Kras G13D is present in 1.24% of AACR GENIE cases, with colon adenocarcinoma, colorectal adenocarcinoma, rectal adenocarcinoma, lung adenocarcinoma, and endometrial endometrioid adenocarcinoma having the greatest prevalence.
Catalog No. NC3787942 Supplier AURORA BIOLABS LLC Supplier No. 57274133GGP100
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. Kras G13D is present in 1.24% of AACR GENIE cases, with colon adenocarcinoma, colorectal adenocarcinoma, rectal adenocarcinoma, lung adenocarcinoma, and endometrial endometrioid adenocarcinoma having the greatest prevalence.