Learn More
Medchemexpress LLC KRAS inhibitor-10 | 2578876-75-0 | 99.9% | 519.63 | 100 MG

Supplier: Medchemexpress LLC HY138295100MG
KRAS inhibitor-10 selectively and effectively inhibits RAS proteins, particularly KRAS proteins. It is an orally active anti-cancer agent and can be used for cancer research, including pancreatic cancer, breast cancer, multiple myeloma, leukemia, and lung cancer.
- Selectively inhibits RAS proteins, particularly KRAS proteins.
- Orally active anti-cancer agent.
- Demonstrates antiproliferative effects in KRas mutant tumor cells.
- Shows significant oral anti-tumor activities in vivo.
- Soluble in DMSO: 100 mg/mL.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.