Learn More
Medchemexpress LLC Kras inhibitor-10 | 2578876-75-0 | 99.9% | 519.63 | 50 MG
SDP
Catalog No. 5000351434
Encompass_Preferred
KRAS inhibitor-10 is a tetrahydroisoquinoline compound that selectively and effectively inhibits RAS proteins, particularly KRAS proteins. It acts as an orally active anti-cancer agent, useful for various cancer research, including studies on pancreatic cancer, breast cancer, multiple myeloma, leukemia, and lung cancer.
  • Selectively and effectively inhibits RAS proteins, particularly KRAS proteins
  • Orally active anti-cancer agent
  • Utilized for cancer research
  • Exhibits antiproliferative effects in KRas mutant tumor cells
  • Shows significant oral anti-tumor activities in vivo
  • Safe and well-tolerated in bearing mice
Catalog No. 50-003-51434 Supplier Medchemexpress LLC Supplier No. HY13829550MG
KRAS inhibitor-10 is a tetrahydroisoquinoline compound that selectively and effectively inhibits RAS proteins, particularly KRAS proteins. It acts as an orally active anti-cancer agent, useful for various cancer research, including studies on pancreatic cancer, breast cancer, multiple myeloma, leukemia, and lung cancer.
  • Selectively and effectively inhibits RAS proteins, particularly KRAS proteins
  • Orally active anti-cancer agent
  • Utilized for cancer research
  • Exhibits antiproliferative effects in KRas mutant tumor cells
  • Shows significant oral anti-tumor activities in vivo
  • Safe and well-tolerated in bearing mice