Learn More
Medchemexpress LLC KRAS inhibitor-9 | 300809-71-6 | 99.9% | 292.81 | 50 MG
SDP
Catalog No. 5000349575
Encompass_Preferred
KRAS inhibitor-9 is a potent inhibitor that blocks the formation of GTP-KRAS and downstream activation of KRAS. It causes G2/M cell cycle arrest and induces apoptosis. This inhibitor selectively inhibits the proliferation of NSCLC cells with KRAS mutation but not normal lung cells.
  • Binds to KRAS G12D, KRAS G12C, and KRAS Q61H protein
  • Induces G2/M cell cycle arrest
  • Promotes apoptosis
  • Selectively inhibits NSCLC cells with KRAS mutation
Catalog No. 50-003-49575 Supplier Medchemexpress LLC Supplier No. HY13749750MG
KRAS inhibitor-9 is a potent inhibitor that blocks the formation of GTP-KRAS and downstream activation of KRAS. It causes G2/M cell cycle arrest and induces apoptosis. This inhibitor selectively inhibits the proliferation of NSCLC cells with KRAS mutation but not normal lung cells.
  • Binds to KRAS G12D, KRAS G12C, and KRAS Q61H protein
  • Induces G2/M cell cycle arrest
  • Promotes apoptosis
  • Selectively inhibits NSCLC cells with KRAS mutation