Learn More
AURORA BIOLABS LLC Kras Q61K–cRAF Binding Assay Kit,100 rxn
Supplier: AURORA BIOLABS LLC 57277615BK100
KRAS is a member of the RAS protein family, a class of small GTPases that regulate important cellular signaling pathways involved in cell growth, differentiation, and survival. KRAS cycles between an inactive GDP-bound state and an active GTP-bound state, which enables interaction with downstream effectors such as cRAF. Mutations in KRAS are commonly associated with human cancers, making the KRAS-RAF signaling pathway an important target for cancer research and drug discovery. The KRAS (Q61K)-cRAF Binding Assay Kit is a TR-FRET–based assay designed to quantitatively detect the interaction between activated KRAS and the cRAF Ras-binding domain (RBD). Activated KRAS generates a measurable FRET signal upon binding to cRAF, while inhibitors that disrupt the interaction reduce the signal intensity, enabling evaluation of potential therapeutic compounds.