Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED KRAS Wild Type Reference Standard 1 FFPE curl, 15 um
Supplier:  HORIZON DISCOVERY BIOSCIENCES LIMITED HD135
Encompass
The KRAS Wild Type Reference Standard is a highly-characterized, biologically-relevant quality control material used to assess the performance of assays that detect somatic mutations, such as Sanger and qPCR sequencing assays.
Catalog No. NC1883047