Learn More
AURORA BIOLABS LLC Kras WT Binding Assay, 384 rxn
Catalog No. NC3786909
Encompass
The Aurora Biolabs KRAS WT c-RAF Binding Assay Kit is a TR-FRET based assay used to quantify binding between active KRAS WT and the Ras Binding Domain (RBD) of c-RAF. The KRAS protein is preloaded with GTP and recognized by a terbium-labeled anti-Tag2 antibody (HTRF donor), while the c-RAF-RBD is labeled with a fluorescent acceptor. Upon binding, FRET occurs and emission at 665nm is measured. This assay provides a robust method for screening inhibitors that block KRAS effector interactions. Ideal for oncology research and drug discovery, each kit contains reagents for 384 reactions.
Catalog No. NC3786909 Supplier AURORA BIOLABS LLC Supplier No. 57274121BK384
The Aurora Biolabs KRAS WT c-RAF Binding Assay Kit is a TR-FRET based assay used to quantify binding between active KRAS WT and the Ras Binding Domain (RBD) of c-RAF. The KRAS protein is preloaded with GTP and recognized by a terbium-labeled anti-Tag2 antibody (HTRF donor), while the c-RAF-RBD is labeled with a fluorescent acceptor. Upon binding, FRET occurs and emission at 665nm is measured. This assay provides a robust method for screening inhibitors that block KRAS effector interactions. Ideal for oncology research and drug discovery, each kit contains reagents for 384 reactions.