Learn More
AURORA BIOLABS LLC Kras WT, GppNHp loaded, 100 μg
Catalog No. NC3787925
Encompass
Kras is a member of the RAS protein family, which are a class of small GTPases involved in cell signaling. Plays a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes (TSGs) in colorectal cancer (CRC) cells in a ZNF304-dependent manner.
Catalog No. NC3787925 Supplier AURORA BIOLABS LLC Supplier No. 5727WTGGP100
Kras is a member of the RAS protein family, which are a class of small GTPases involved in cell signaling. Plays a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes (TSGs) in colorectal cancer (CRC) cells in a ZNF304-dependent manner.