Learn More
Medchemexpress LLC Kras4B protein, human (His) | 97.0% | 50UG
SDP
Catalog No. 5000175589
Encompass_Preferred
Recombinant human KRAS4B protein (isoform 2) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized. The construct spans residues M1-K169 (169 amino acids), with an approximate molecular weight of 23.0 kDa. Designed for research use in biochemical assays, binding studies, and structural analysis.
  • N-terminal 6xHis affinity tag for purification and detection.
  • expressed in E. coli and supplied lyophilized for long-term storage.
  • 169 amino acids, approx. 23.0 kDa molecular weight.
  • purity greater than 95% by reducing SDS-PAGE (certificate of analysis result 97.0%).
  • formulated with 20 mM PB, 100 mM NaCl, 8% trehalose, 4% mannitol, 0.02% tween 80, pH 7.5.
  • suitable for biochemical assays, binding studies, and structural analysis.
  • validated by certificate of analysis for purity and identity.
Catalog No. 50-001-75589 Supplier Medchemexpress LLC Supplier No. HYP70233A50UG
Recombinant human KRAS4B protein (isoform 2) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized. The construct spans residues M1-K169 (169 amino acids), with an approximate molecular weight of 23.0 kDa. Designed for research use in biochemical assays, binding studies, and structural analysis.
  • N-terminal 6xHis affinity tag for purification and detection.
  • expressed in E. coli and supplied lyophilized for long-term storage.
  • 169 amino acids, approx. 23.0 kDa molecular weight.
  • purity greater than 95% by reducing SDS-PAGE (certificate of analysis result 97.0%).
  • formulated with 20 mM PB, 100 mM NaCl, 8% trehalose, 4% mannitol, 0.02% tween 80, pH 7.5.
  • suitable for biochemical assays, binding studies, and structural analysis.
  • validated by certificate of analysis for purity and identity.