Learn More
Cayman Chemical Kratom AlkaloId MIxture 1ea
SDP
Catalog No. 502915884
Encompass_Preferred
A mixture containing alkaloids found in M. speciosa (Kratom) in an ammonia in methanol solution; designed for use in methods development and as a reference standard for these substances during routine analytical checks using HPLC or LC-MS instrumentation; intended for research and forensic applications
Catalog No. 50-291-5884 Supplier Cayman Chemical Supplier No. 376491 EA
A mixture containing alkaloids found in M. speciosa (Kratom) in an ammonia in methanol solution; designed for use in methods development and as a reference standard for these substances during routine analytical checks using HPLC or LC-MS instrumentation; intended for research and forensic applications