Learn More
ABclonal Technology KRBOX4 Rabbit pAb
SDP
Catalog No. 5000107152
Encompass_Preferred
This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked cognitive disability. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07152 Supplier ABclonal Technology Supplier No. A14514500UL
This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked cognitive disability. Multiple transcript variants encoding different isoforms have been found for this gene.