Learn More
TARGETMOL CHEMICALS INC KRCA-0008 25MG
SDP
Catalog No. 502878231
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KRCA-0008 is an effective and specific ALK/Ack1 inhibitor (IC50 12/4 nM); displays drug-like properties without hERG liability. Purity 96.19%
Catalog No. 50-287-8231 Supplier TARGETMOL CHEMICALS INC Supplier No. T264125MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KRCA-0008 is an effective and specific ALK/Ack1 inhibitor (IC50 12/4 nM); displays drug-like properties without hERG liability. Purity 96.19%