Learn More
Aviva Systems Biology Corp KRCC1 Antibody - N-terminal region (ARP56951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRCC1. It was validated on Western Blot. KRCC1,lysine-rich coiled-coil 1,CHBP2; FLJ22333; KRCC1,lysine-rich coiled-coil protein 1,lysine-rich coiled-coil protein 1,cryptogenic hepatitis binding protein,cryptogenic hepatitis-binding protein 2The immunogen is a synthetic peptide directed towards the N terminal region of human KRCC1. Alternate names: CHBP2, FLJ22333ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-286-242