Learn More
Cayman Chemical Krebs Cycle stnd MIXure 100mL
SDP
Catalog No. 502941058
Encompass_Preferred
A mixture of seven non-volatile acid Krebs cycle intermediates; intended for use as a reference standard for the detection of these intermediates in various sample types by GC-MS
Catalog No. 50-294-1058 Supplier Cayman Chemical Supplier No. 28681100 ML
A mixture of seven non-volatile acid Krebs cycle intermediates; intended for use as a reference standard for the detection of these intermediates in various sample types by GC-MS