Learn More
Teknova Krebs-Henseleit Buffer. 1000mL, Sterile.
SDP
Catalog No. 502388116
Encompass
0.200% Cerelose Anhydrous Dextrose (Glucose), 0.014% Magnesium Sulfate, 0.016% Monopotassium Phosphate, 0.035% Potassium Chloride, 0.690% Sodium Chloride, 373g/mL Calcium Chloride, 0.210% Sodium Carbonate
Catalog No. 50-238-8116 Supplier Teknova Supplier No. K2352
0.200% Cerelose Anhydrous Dextrose (Glucose), 0.014% Magnesium Sulfate, 0.016% Monopotassium Phosphate, 0.035% Potassium Chloride, 0.690% Sodium Chloride, 373g/mL Calcium Chloride, 0.210% Sodium Carbonate