Learn More
Sigma Aldrich Fine Chemicals Biosciences Krebs-Henseleit Buffer Modified With 2000 mg/L glucose, without calcium chloride and sodium bicarbonate, powder, suitable for cell culture | 10X1L
Catalog No. 501657167
Encompass
Krebs-Henseleit Buffer Modified With 2000 mg/L glucose, without calcium chloride and sodium bicarbonate, powder, suitable for cell culture | 10X1L
Catalog No. 50-165-7167 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. K375310X1L
Krebs-Henseleit Buffer Modified With 2000 mg/L glucose, without calcium chloride and sodium bicarbonate, powder, suitable for cell culture | 10X1L