Learn More
Sigma Aldrich Fine Chemicals Biosciences Krebs-Ringer Bicarbonate Buffer, 10 x 1 L
Catalog No. NC0936430
Encompass
Bicarbonate buffered isotonic saline containing magnesium, potassium, sodium and phosphate ions.
Catalog No. NC0936430 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. K400210X1L
Bicarbonate buffered isotonic saline containing magnesium, potassium, sodium and phosphate ions.