Learn More
Crescent Chemical Co Inc KRESOXIM-METHYL 100 MG
SDP
Catalog No. 5074331
Encompass_Preferred
Kresoxim-methyl Fungicide (WS) CAS 143390-89-0 Molecular Weight - 313.3478 Storage - +5°C Catalog Num - DRE-C14570000 pesticide
Catalog No. 50-743-31 Supplier Crescent Chemical Co Inc Supplier No. PSC14570000
Kresoxim-methyl Fungicide (WS) CAS 143390-89-0 Molecular Weight - 313.3478 Storage - +5°C Catalog Num - DRE-C14570000 pesticide