Learn More
Cayman Chemical KresoxIm-methyl 100mg
SDP
Catalog No. 502932855
Encompass_Preferred
A fungicide; inhibits conidial germination of V. inaequalis isolates from apple orchards (EC50s = 0.00033-0.0078 mg/L); inhibits mycelial growth (EC50 = 0.24 mg/L) and is fungicidal against Saprolegnia (MIC = 1 mg/L); increases intracellular calcium levels and disrupts the mitochondrial membrane potential in mouse cortical cultures; toxic to goldfish (LC50 = 0.807 mg/L)
Catalog No. 50-293-2855 Supplier Cayman Chemical Supplier No. 25816100 MG
A fungicide; inhibits conidial germination of V. inaequalis isolates from apple orchards (EC50s = 0.00033-0.0078 mg/L); inhibits mycelial growth (EC50 = 0.24 mg/L) and is fungicidal against Saprolegnia (MIC = 1 mg/L); increases intracellular calcium levels and disrupts the mitochondrial membrane potential in mouse cortical cultures; toxic to goldfish (LC50 = 0.807 mg/L)