Learn More
Cayman Chemical Kresoxim-methyl, Purity: ≥95%, CAS: 143390-89-0, 50 mg
SDP
Catalog No. 502936812
Encompass_Preferred
A fungicide; inhibits conidial germination of V. inaequalis isolates from apple orchards (EC50s = 0.00033-0.0078 mg/L); inhibits mycelial growth (EC50 = 0.24 mg/L) and is fungicidal against Saprolegnia (MIC = 1 mg/L); increases intracellular calcium levels and disrupts the mitochondrial membrane potential in mouse cortical cultures; toxic to goldfish (LC50 = 0.807 mg/L)
Catalog No. 50-293-6812 Supplier Cayman Chemical Supplier No. 2581650 MG
A fungicide; inhibits conidial germination of V. inaequalis isolates from apple orchards (EC50s = 0.00033-0.0078 mg/L); inhibits mycelial growth (EC50 = 0.24 mg/L) and is fungicidal against Saprolegnia (MIC = 1 mg/L); increases intracellular calcium levels and disrupts the mitochondrial membrane potential in mouse cortical cultures; toxic to goldfish (LC50 = 0.807 mg/L)