Learn More
Med Vet International Krey Hook, 8in, J0019, Each
Catalog No. 5000585378
Encompass_Preferred
This double-action OB hook will bite into part of the fetus in the process of doing an embryotomy. Should the instrument slip, the hooks will close automatically and prevent injury.Stainless steel. 8"L.
Catalog No. 50-005-85378 Supplier Med Vet International Supplier No. JOR19
This double-action OB hook will bite into part of the fetus in the process of doing an embryotomy. Should the instrument slip, the hooks will close automatically and prevent injury.Stainless steel. 8"L.