Learn More
Medchemexpress LLC KRFK TFA | 99.8% | 577.73 | 25 MG
SDP
Catalog No. 5000397415
Encompass_Preferred
KRFK TFA is a peptide derived from TSP-1 that can activate TGF-β. It promotes TGF-β-mediated signaling and its downstream role, independent of thrombospondin (TSP) receptors such as CD47 and CD36. This product can be used for research on chronic ocular surface inflammatory disorders.
  • Activates TGF-β
  • Promotes TGF-β-mediated signaling
  • Independent of thrombospondin (TSP) receptors
  • Suitable for research on chronic ocular surface inflammatory disorders
Catalog No. 50-003-97415 Supplier Medchemexpress LLC Supplier No. 25MGHYP3970A
KRFK TFA is a peptide derived from TSP-1 that can activate TGF-β. It promotes TGF-β-mediated signaling and its downstream role, independent of thrombospondin (TSP) receptors such as CD47 and CD36. This product can be used for research on chronic ocular surface inflammatory disorders.
  • Activates TGF-β
  • Promotes TGF-β-mediated signaling
  • Independent of thrombospondin (TSP) receptors
  • Suitable for research on chronic ocular surface inflammatory disorders