Learn More
Selleck Chemical LLC KRIBB11 10mg 342639-96-7
SDP
Supplier:  Selleck Chemical LLC S840210MG
Encompass_Preferred
KRIBB11 abolishes the heat shock-induced luciferase activity with an IC50 of 1.2 ?M. It is an inhibitor of the transcription factor Heat Shock Factor 1 (HSF1). KRIBB11 induces growth arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6029