Learn More
Medchemexpress LLC HY-100872 5mg Medchemexpress, KRIBB11 CAS:342639-96-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008725MG
Encompass_Preferred
Medchemexpress, HY-100872 5mg KRIBB11 CAS:342639-96-7 KRIBB11 is an inhibitor of Heat shock factor 1 (HSF1) , with IC 50 of 1.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1979