Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative measurement of CHO Host Cell Proteins as a contaminant in various biological preparations. Assay Range: 0.312 - 20 ng/ml; Sensitivity: 0.188 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBBP03
Encompass_Preferred
Pack Size: 1 x 96 wells.This CHO HCP ELISA detects CHO Host Cell Protein contamination accurately and consistently. Features include validation against leading brands, long shelf life, and minimal lot-to-lot consistency.Introduction: A variety of proteins which are used as therapeutic agents in humans and animals are produced through recombinant expression in Chinese Hamster Ovary (CHO) cells. The manufacturing and purification process of these products tends to leave the potential for contamination by Host Cell Proteins (HCPs) from CHO cells, which may result in adverse toxic or immunological reactions that ultimately affect the efficacy of the therapeutic agent. The simple, objective and semi-quantitative ELISA is a highly sensitive method that aids in purification process development, process control, quality control and product release testing optimally.
Catalog No. NC2747129