Learn More
KRISHGEN BIOSYSTEMS Immunoassay for quantitative estimation of Semaglutide in human serum and plasma. Validated against OZEMPIC and local biosimilars for PK studies. Assay Range: 50 - 3000 ng/ml; Sensitivity: 50 ng/ml
Supplier:  KRISHGEN BIOSYSTEMS KBI5030
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: Semaglutide (trade name Ozempic) is a pharmaceutical drug in development by a Danish company Novo Nordisk for the treatment of type 2 diabetes. It is marketed by the name Ozempic. As a glucagon-like peptide-1 receptor agonist, it lowers the blood sugar level by increasing the production of insulin. It was discovered in 2012, by a team of researchers at Novo Nordisk as a longer-acting alternative to liraglutide. This Semaglutide ELISA accurately quantifies semaglutide and biosimilars in human samples. If you are looking to use animal samples, please contact us. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <10%.
Catalog No. NC2747080