Learn More
TARGETMOL CHEMICALS INC KRM-III 5MG
SDP
Catalog No. 502576909
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KRM-III is an orally active T cell antigen receptor (TCR) inhibitor with anti-inflammatory activity that potently inhibits TCR and myristate acetate/fosfomycin/ionomycin-induced NFAT nuclear factor activation and T cell proliferation with an IC50 of approximately 5 uM. purity: 99%
Catalog No. 50-257-6909 Supplier TARGETMOL CHEMICALS INC Supplier No. T358855MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KRM-III is an orally active T cell antigen receptor (TCR) inhibitor with anti-inflammatory activity that potently inhibits TCR and myristate acetate/fosfomycin/ionomycin-induced NFAT nuclear factor activation and T cell proliferation with an IC50 of approximately 5 uM. purity: 99%