Learn More
Medchemexpress LLC KRM-III | 79220-94-3 | 98.1% | 252.33 | 1 ML
SDP
Catalog No. 5000349662
Encompass_Preferred
KRM-III is a potent and orally active T-cell antigen receptor (TCR) inhibitor. It inhibits TCR- and phorbol myristate acetate/ionomycin-induced activation of nuclear factor of activated T cells (NFAT) and T-cell proliferation with an IC50 of approximately 5 μM. This product also exhibits anti-inflammatory activity and is intended for research use only.
  • Potent and orally active T-cell antigen receptor (TCR) inhibitor.
  • Inhibits TCR- and phorbol myristate acetate/ionomycin-induced activation of nuclear factor of activated T cells (NFAT).
  • Inhibits T-cell proliferation with an IC50 of approximately 5 μM.
  • Exhibits anti-inflammatory activity.
Catalog No. 50-003-49662 Supplier Medchemexpress LLC Supplier No. HY13642710MM/1ML
KRM-III is a potent and orally active T-cell antigen receptor (TCR) inhibitor. It inhibits TCR- and phorbol myristate acetate/ionomycin-induced activation of nuclear factor of activated T cells (NFAT) and T-cell proliferation with an IC50 of approximately 5 μM. This product also exhibits anti-inflammatory activity and is intended for research use only.
  • Potent and orally active T-cell antigen receptor (TCR) inhibitor.
  • Inhibits TCR- and phorbol myristate acetate/ionomycin-induced activation of nuclear factor of activated T cells (NFAT).
  • Inhibits T-cell proliferation with an IC50 of approximately 5 μM.
  • Exhibits anti-inflammatory activity.