Learn More
Medchemexpress LLC KRN2 bromide | 1390654-28-0 | 98.8% | 524.38 | 25 MG
SDP
Catalog No. 5000439833
Encompass_Preferred
KRN2 bromide is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. It selectively suppresses the expression of pro-inflammatory genes, including Nos2 and Il6. It also dose-dependently inhibits NF-κB p65 binding to Nfat5 promoter 1 and directly blocks the interaction between NF-κB p65 and its DNA binding sequence.
  • Selectively inhibits nuclear factor of activated T cells (NFAT5).
  • Suppresses expression of pro-inflammatory genes.
  • Inhibits NF-κB p65 binding to Nfat5 promoter.
  • Effectively suppresses AIA and CIA in mice, decreasing pro-inflammatory cytokines and autoantibodies.
Catalog No. 50-004-39833 Supplier Medchemexpress LLC Supplier No. 25MGHY112125A
Add to Cart
Add to Cart
KRN2 bromide is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. It selectively suppresses the expression of pro-inflammatory genes, including Nos2 and Il6. It also dose-dependently inhibits NF-κB p65 binding to Nfat5 promoter 1 and directly blocks the interaction between NF-κB p65 and its DNA binding sequence.
  • Selectively inhibits nuclear factor of activated T cells (NFAT5).
  • Suppresses expression of pro-inflammatory genes.
  • Inhibits NF-κB p65 binding to Nfat5 promoter.
  • Effectively suppresses AIA and CIA in mice, decreasing pro-inflammatory cytokines and autoantibodies.