Learn More
TARGETMOL CHEMICALS INC KRN2 BROMIDE 5MG
SDP
Catalog No. 502577445
Encompass_Preferred
Also available in 2mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. KRN2 bromide is a potent selective inhibitor of nuclear factor of activated T cells (NFAT5 IC50= 0.1 μM). purity: 99%
Catalog No. 50-257-7445 Supplier TARGETMOL CHEMICALS INC Supplier No. T117825MG
Add to Cart
Add to Cart
Also available in 2mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. KRN2 bromide is a potent selective inhibitor of nuclear factor of activated T cells (NFAT5 IC50= 0.1 μM). purity: 99%