Learn More
Medchemexpress LLC HY-112125A 5mg Medchemexpress, KRN2 (bromide) CAS:1390654-28-0 Purity:>98%
SDP
Catalog No. 501969028
Encompass_Preferred
Medchemexpress, HY-112125A 5mg KRN2 (bromide) CAS:1390654-28-0 KRN2 (bromide) is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9028 Supplier Medchemexpress LLC Supplier No. HY112125A5MG
Medchemexpress, HY-112125A 5mg KRN2 (bromide) CAS:1390654-28-0 KRN2 (bromide) is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.