Learn More
Medchemexpress LLC HY-112126 5mg Medchemexpress, KRN5 CAS:1800465-47-7 Purity:>98%
SDP
Catalog No. 501969029
Encompass_Preferred
Medchemexpress, HY-112126 5mg KRN5 CAS:1800465-47-7 KRN5, a derivative of KRN2, is an oral active Nuclear factor of activated T cells 5 (NFAT5) suppressor, with an IC50 of 750 nM. KRN5 has potential to treat NFAT5-mediated Chronic Arthritis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9029 Supplier Medchemexpress LLC Supplier No. HY1121265MG
Medchemexpress, HY-112126 5mg KRN5 CAS:1800465-47-7 KRN5, a derivative of KRN2, is an oral active Nuclear factor of activated T cells 5 (NFAT5) suppressor, with an IC50 of 750 nM. KRN5 has potential to treat NFAT5-mediated Chronic Arthritis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.