Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kroll’s Reagent Meets ASTM E407 Specification Etchant 192, Reagents

Catalog No. 01334888
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-888 100 mL
01-334-886 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-888 Supplier Reagents Holdings Llc Supplier No. CK010500100A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kroll’s Reagent Meets ASTM E407 Specification Etchant 192, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Colorless to Slight Yellow
Boiling Point 100°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.19
pH <1
Quantity 100 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Kroll’s Reagent Meets ASTM E407 Specification Etchant 192
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.