Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kroll’s Reagent Meets ASTM E407 Specification Etchant 192, Reagents

Kroll’s Reagent Meets ASTM E407 Specification Etchant 192, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$106.20 - $436.00

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-888
Accessories View Documents
Reagents Holdings Llc
CK010500100A
100 mL
Each for $106.20
Add to Cart
 
 Restricted Item
01-334-887
Accessories View Documents
Reagents Holdings Llc
CK0105001A
1 L
Each for $436.00
Add to Cart
 
 Restricted Item
01-334-886
Accessories View Documents
Reagents Holdings Llc
CK010500250A
250 mL
Each for $239.50
Add to Cart
 
 Restricted Item
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7697-37-2
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless to Slight Yellow
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.19
Non-sterile
Kroll’s Reagent Meets ASTM E407 Specification Etchant 192
7732-18-5 , 7697-37-2
H2O
O
18.02
Poly Bottle
<1
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.