Learn More
Sigma Aldrich Fine Chemicals Biosciences Kromasil(R) C18 HPLC Column 5 mum particle size, pore size 100 Ã…, L x I.D. 150 mm x 4.6 mm |
Catalog No. NC0930806
Encompass
Kromasil(R) C18 HPLC Column 5 mum particle size, pore size 100 Ã…, L x I.D. 150 mm x 4.6 mm |
Catalog No. NC0930806 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. K08670356
Kromasil(R) C18 HPLC Column 5 mum particle size, pore size 100 Ã…, L x I.D. 150 mm x 4.6 mm |