Learn More
MSC Kromer Welding Cap, Blue, All-Cotton, Double-Stitched, 1 ea
Catalog No. 5000677548
Encompass
This American-made welding cap is designed for safety and comfort, featuring an all-cotton construction with a spacious balloon top and sweatband. Each cap undergoes strict quality control and individual inspection, ensuring durability and consistent performance.
  • All-cotton balloon top is spacious and comfortable.
  • Cotton sweatband keeps perspiration out of eyes.
  • Produced under strict quality control standards.
  • Each cap is individually inspected by skilled union craftsmen.
  • Meticulously constructed with tough double-stitching.
  • Available in sizes from 6-7/8 up to 7-7/8.
  • Made in the USA.
  • Intended to be worn under a welding hood, not in place of one.
Catalog No. 50-006-77548 Supplier MSC Supplier No. 40604894
This American-made welding cap is designed for safety and comfort, featuring an all-cotton construction with a spacious balloon top and sweatband. Each cap undergoes strict quality control and individual inspection, ensuring durability and consistent performance.
  • All-cotton balloon top is spacious and comfortable.
  • Cotton sweatband keeps perspiration out of eyes.
  • Produced under strict quality control standards.
  • Each cap is individually inspected by skilled union craftsmen.
  • Meticulously constructed with tough double-stitching.
  • Available in sizes from 6-7/8 up to 7-7/8.
  • Made in the USA.
  • Intended to be worn under a welding hood, not in place of one.