Learn More
REPLIGEN CORPORATION Repligen KrosFlo K04-E750-10-N, 750 kD MWCO, mPES Membrane, 1.00 mm Fiber ID
SDP
Supplier:  REPLIGEN CORPORATION K04E75010N
Encompass
The Repligen KrosFlo K04-E750-10-N is a high-performance hollow fiber filter module designed for efficient ultrafiltration and diafiltration. It features a 750 kD mPES membrane that minimizes fouling and maximizes recovery, making it ideal for shear-sensitive applications like lentivirus and mRNA processing. With a robust design for high-pressure use, this filter module ensures gentle handling of biologics, setting it apart in laboratory filtration solutions.
Catalog No. NC3716969