Learn More
Medchemexpress LLC KRpep-2d TFA | 2098181-84-9 | 95.1% | 2675.03 | 2 MG
SDP
Catalog No. 5000397438
Encompass_Preferred
KRpep-2d is a potent inhibitor of K-Ras, designed to impede the proliferation of K-Ras-driven cancer cells. It is suitable for use in cancer research. The product possesses a cyclic structure, which is crucial for its K-Ras inhibitory activity, with specific amino acid residues (Leu77, Ile9, and Asp12) being critical for this function.
  • Potently inhibits K-Ras activity.
  • Reduces the proliferation of cancer cells driven by K-Ras.
  • Features a cyclic structure important for its inhibitory action.
  • Critical amino acid residues contribute to its effectiveness.
  • Can be used for cancer research applications.
Catalog No. 50-003-97438 Supplier Medchemexpress LLC Supplier No. HYP3277A2MG
KRpep-2d is a potent inhibitor of K-Ras, designed to impede the proliferation of K-Ras-driven cancer cells. It is suitable for use in cancer research. The product possesses a cyclic structure, which is crucial for its K-Ras inhibitory activity, with specific amino acid residues (Leu77, Ile9, and Asp12) being critical for this function.
  • Potently inhibits K-Ras activity.
  • Reduces the proliferation of cancer cells driven by K-Ras.
  • Features a cyclic structure important for its inhibitory action.
  • Critical amino acid residues contribute to its effectiveness.
  • Can be used for cancer research applications.