Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRpTIRR acetate | 98.2% | 908.99 | 1 MG
SDP

Catalog No. 5000398009
Encompass_Preferred

KRpTIRR acetate is a phosphothreonine-containing peptide that serves as a substrate for phosphatases.

  • Appearance: Solid, white to off-white
  • Sequence: Lys-Arg-{Thr}-Ile-Arg-Arg (KR-{Thr}-IRR)
  • Solubility in water: Greater than or equal to 100 mg/mL
  • Functions as a peptide, enzyme substrate, and phosphatase inhibitor

Catalog No. 50-003-98009 Supplier Medchemexpress LLC Supplier No. HYP5441A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRpTIRR acetate is a phosphothreonine-containing peptide that serves as a substrate for phosphatases.

  • Appearance: Solid, white to off-white
  • Sequence: Lys-Arg-{Thr}-Ile-Arg-Arg (KR-{Thr}-IRR)
  • Solubility in water: Greater than or equal to 100 mg/mL
  • Functions as a peptide, enzyme substrate, and phosphatase inhibitor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.