Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRpTIRR acetate | 98.2% | 908.99 (free base) | 5 MG
SDP

Catalog No. 5000398022
Encompass_Preferred

KRpTIRR acetate is a phosphothreonine-containing peptide that functions as a substrate for phosphatases. It is intended for research use only.

  • Purity: 98.24%
  • Form: Solid
  • Color: White to off-white
  • Sequence: Lys-Arg-{Thr}-Ile-Arg-Arg
  • Shortened sequence: KR-{Thr}-IRR
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -80°C for 2 years, -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): ≥ 100 mg/mL in H2O

Catalog No. 50-003-98022 Supplier Medchemexpress LLC Supplier No. HYP5441A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRpTIRR acetate is a phosphothreonine-containing peptide that functions as a substrate for phosphatases. It is intended for research use only.

  • Purity: 98.24%
  • Form: Solid
  • Color: White to off-white
  • Sequence: Lys-Arg-{Thr}-Ile-Arg-Arg
  • Shortened sequence: KR-{Thr}-IRR
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -80°C for 2 years, -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): ≥ 100 mg/mL in H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.