Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3858MSM6P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: KRT10/844 + KRT10/1275. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 s kappa. Positive Control: A431, HeLa, MCF7 cells. Esophagus or tonsil. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human KRT10 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 56.5kDa.Chromosome: 17q21.2. Gene Symbol: KRT10. Gene ID: 3858. Ensemble: ENSG00000186395. Uniprot: NP_000412.
Catalog No. 50-188-4883