Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM11X606P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM262. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: A431, HeLa, MCF7 cells. Tonsil, esophageal or bladder carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 1.0mg/ml. Purity: Protein A/G Purified. Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 56.5kDa (CK10) & 53kDa (CK13).Chromosome: 17q21.2. Gene Symbol: KRT10. Gene ID: 3858. Ensemble: ENSG00000186395. Uniprot: NP_000412.
Catalog No. 50-188-5406