Learn More
Aviva Systems Biology Corp KRT10 Antibody - N-terminal region (ARP41730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KRT10. Alternate names: CK10, K10, KPP, BIE, EHK, BCIEApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57971